Anti-IgG (H+L) Mouse Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverantör: Rockland Immunochemicals
611-300-122
ROCK611-300-122EA 4900 SEK
ROCK611-300-122
Anti-IgG (H+L) Mouse Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikroppar
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Mouse
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR