Anti-IgM µ Rabbit Polyclonal Antibody (Peroxidase)

Leverantör: Rockland Immunochemicals
610-4307
ROCK610-4307EA 5200 SEK
ROCK610-4307
Anti-IgM µ Rabbit Polyclonal Antibody (Peroxidase)
Antikroppar
Mouse IgM (mu chain) peroxidase conjugated antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays.

Recommended Dilutions: ELISA: 1:180,000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR