Anti-IgM µ Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverantör: Rockland Immunochemicals
609-1007
ROCK609-1007EA 3075 SEK
ROCK609-1007
Anti-IgM µ Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikroppar
Secondary Goat Anti-IgM (mu chain) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR