Anti-GST Goat Polyclonal Antibody (Biotin)

Leverantör: Rockland Immunochemicals
600-106-200
ROCK600-106-200EA 13975 SEK
ROCK600-106-200
Anti-GST Goat Polyclonal Antibody (Biotin)
Antikroppar
Primary Goat Anti-GLUTATHIONE-S-TRANSFERASE (Schistosoma japonicum) IgG Fraction Reacts with Pan

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR