Anti-NEDD9 Mouse Monoclonal Antibody (Unconjugated) [clone: 14A11]

Leverantör: Rockland Immunochemicals
200-301-912
ROCK200-301-912EA 9775 SEK
ROCK200-301-912
Anti-NEDD9 Mouse Monoclonal Antibody (Unconjugated) [clone: 14A11]
Antikroppar
This monoclonal antibody has been tested for use in western blotting, immunoprecipitation and immunofluorescence.  This clone recognizes HEF1 under non-denaturing conditions.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Immunofluorescence: 1:100; Western Blot: 1:500; Immunoprecipitation: 1:100;

Type: Primary
Antigen: NEDD9
Clonality: Monoclonal
Clone: 14A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR