Anti-ALB Rabbit Polyclonal Antibody

Leverantör: Rockland Immunochemicals
109-4133
ROCK109-4133EA 5850 SEK
ROCK109-4133
Anti-ALB Rabbit Polyclonal Antibody
Antikroppar
Anti-ALB antibody is suitable for western blotting. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum, purified human serum albumin IgG, and human Serum.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ALB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFIKATIONER

Antigensymbol ALB
Antikroppsnamn albumin
Environmentally Preferable
Antigen synonymer PRO1341|PRO0883|ANALBA|FDAH|PRO0903
Antikroppstyp Primär
Klonalitet Polyclonal
Konjugation Unconjugated
Reaktivitet Human
Värd Rabbit
Gen-ID 213
Isotyp
Western blot Yes
ELISA Yes
Immunutfällning Yes
Immunogen Albumin (Human Serum)
Mått Antiserum
Form Lyophilized
Rening delipidation and defibrination
Koncentration 80 mg/ml by Refractometry
Korsad adsorption none
Storage buffer 0,01% (w/v) Sodium Azide,
Förvaringstemperatur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR