Anti-MGRN1 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
25-831
PRSI25-831EA 7675 SEK
PRSI25-831
Anti-MGRN1 Rabbit Polyclonal Antibody
Antikroppar
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MGRN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

SPECIFIKATIONER

Antigensymbol MGRN1
Antikroppsnamn Mahogunin RING Finger Protein 1
Environmentally Preferable
Antigen synonymer KIAA0544|RING Finger Protein 156|Mahogunin|Ring Finger 1|RNF156
Klonalitet Polyclonal
Konjugation Unconjugated
Reaktivitet Human,Rat,Mouse
Värd Rabbit
Isotyp
Western blot Yes
ELISA Yes
Immunogen A synthetic peptide corresponding a region of human MGRN1.
Form Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR