Pierce™ Digestion Indicator for Mass Spectrometry, Thermo Scientific
Leverantör: Thermo Fisher Scientific
84841
PIER84841EA 4675 SEK
PIER84841
Pierce™ Digestion Indicator for Mass Spectrometry, Thermo Scientific
Proteinextraktions Reagens för proteinextraktion
The Digestion Indicator for Mass Spectrometry is a non-mammalian recombinant protein (26kDa) with five signature peptides for use in determining the digestion efficiency and reproducibility across multiple samples.
  • Versatile
  • For use with mass spectrometer
  • Non-mammalian protein source
This product is sufficient for production of five signature peptides upon digestion for mass spectrometry.
Observera: For Research Use Only. Not for use in diagnostic procedures.