Dissolved oxygen meters, bench, Orion™ Versa Star Pro™
Leverantör: Thermo Orion
VSTAR30 VSTAR32
ORIOVSTAR30EA 36250 SEK
ORIOVSTAR30 ORIOVSTAR32
Dissolved oxygen meters, bench, Orion™ Versa Star Pro™
Elektrokemimätare Syremätare
These bench top meters with dissolved oxygen / RDO module meet most challenging applications for dissolved oxygen and temperature. The meters offer interchangeable measurement modules that allow multiple users to customise four separate channels to meet their specific requirements.
  • Can be used with RDO optical or polarographic DO probes
  • Colour display with adjustable text sizes; numeric keypad; onscreen measurement stability indicator
  • Auto-Read to hold a stable reading; ten password-protected methods per channel
  • Audible alarms automatically notify users
More Product Information