ATC probe, Orion™
Leverantör: Thermo Orion
917001
ORIO917001EA 14450 SEK
ORIO917001
ATC probe, Orion™
Elektroder pH-elektroder
ATC probe for general purpose applications. For aqueous samples, intermittent use in methanol or ethanol.
  • With dual Banana plug
  • Durable thanks to epoxy body
  • For use with older Orion meters