pH electrode, Orion™ 9120APWP KNIpHE™
Leverantör: Thermo Orion
9121APWP
ORIO9121APWPEA 3775 SEK
ORIO9121APWP
pH electrode, Orion™ 9120APWP KNIpHE™
Elektroder pH-elektroder
This speciality electrode is ideal for performing accurate determinations of pH in meat matrices.
  • Surrounding stainless steel cutting blade eliminates breakage
  • Ag/AgCl double junction
Leveransinformation: Supplied with 60 ml bottle of electrode fill solution.