Anti-SCN2A Rabbit Polyclonal Antibody

Leverantör: OriGene
TA322241
ORIGTA322241EA 6150 SEK
ORIGTA322241
Anti-SCN2A Rabbit Polyclonal Antibody
Antikroppar
Recommended Dilutions: ELISA: 1:1000-5000; Immunochemistry: 1:25-100

Type: Primary
Antigen: SCN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR