Anti-CUEDC2 Rabbit Polyclonal Antibody

Leverantör: OriGene
TA322189
ORIGTA322189EA 6150 SEK
ORIGTA322189
Anti-CUEDC2 Rabbit Polyclonal Antibody
Antikroppar
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: CUEDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR