Anti-BFSP2 Mouse Monoclonal Antibody [clone: 1A11]

Leverantör: OriGene
CF804391
ORIGCF804391EA 9675 SEK
ORIGCF804391
Anti-BFSP2 Mouse Monoclonal Antibody [clone: 1A11]
Antikroppar
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: BFSP2
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR