Anti-ACOT12 Mouse Monoclonal Antibody [clone: 1A11]

Leverantör: OriGene
CF502396
ORIGCF502396EA 9675 SEK
ORIGCF502396
Anti-ACOT12 Mouse Monoclonal Antibody [clone: 1A11]
Antikroppar
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ACOT12
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR