Anti-ZIC1 Rabbit Polyclonal Antibody (Biotin)

Leverantör: Novus Biologicals
NB600-488B
NOVUNB600-488BEA 8250 SEK
NOVUNB600-488B
Anti-ZIC1 Rabbit Polyclonal Antibody (Biotin)
Antikroppar
Type: Primary
Antigen: ZIC1 (Zic family member 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR