Mag-Bind® RXNPure Plus kit
Leverantör: OMEGA BIO-TEK
M1386-01
M1386-01EA 9100 SEK
M1386-01
Mag-Bind® RXNPure Plus kit
Nukleinsyrareagens Nucleic Acid Purification Kits and Reagents
Mag-Bind® RXNPure Plus Kit allows rapid and reliable isolation DNA from PCR and enzymatic reactions with high recovery rates. The system combines proprietary chemistries with the reversible nucleic acid-binding properties of magnetic beads that selectively bind PCR amplicons 100 bp and larger, and eliminate excess nucleotides, primers and small, non targeted amplification products, such as primer dimers.
  • Efficiently removes excess primers, primer-dimers, dNTPs and salts
  • No centrifugation/filtration steps
  • Scalable - can be adapted to most standard liquid handling robots
  • Use in 96- or 384- well format
This kit is designed for both manual and fully automated purification and may not require reprogramming of liquid handling instruments depending on your current method.