Anti-kappa light chains Antibody (HRP (Horseradish Peroxidase))

Leverantör: MP Biomedicals
0855233
ICNA0855233EA 7125 SEK
ICNA0855233
Anti-kappa light chains Antibody (HRP (Horseradish Peroxidase))
Antikroppar
Peroxidase-Conjugated Goat IgG Fraction To Human Kappa Chain (Bound) is suitable for use as a reagent in enzyme immunoassays (EIA), cell and immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections, cell and tissue labeling (for electron microscopy), and blot immunostaining.

Type: Secondary
Antigen: Kappa light chains
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR