Anti-CASP11 Rabbit Polyclonal Antibody

Leverantör: ENZO LIFE SCIENCES
BMLSA3240100
ENZOBMLSA3240100EA 5025 SEK
ENZOBMLSA3240100
Anti-CASP11 Rabbit Polyclonal Antibody
Antikroppar
Host: Rabbit

  • For Western Blot

Recognizes mouse caspase-11.

Type: Primary
Antigen: CASP11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR