Anti-ESR1 Rabbit Polyclonal Antibody

Leverantör: ENZO LIFE SCIENCES
ALX210201C050
ENZOALX210201C050EA 10100 SEK
ENZOALX210201C050
Anti-ESR1 Rabbit Polyclonal Antibody
Antikroppar
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human and rat endogenous ERalpha . Detects a band of ~64kDa by Western blot. Does not cross-react with ERbeta.

Type: Primary
Antigen: ESR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR