Anti-ELL3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Leverantör: Bioss
BS-8560R-A750
BOSSBS-8560R-A750EA 10375 SEK
BOSSBS-8560R-A750
Anti-ELL3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikroppar
Eukaryotic RNA polymerase II mediates the synthesis of mature and functional messenger RNA. This is a multistep process, called the transcription cycle, that includes five stages: preinitiation, promoter, clearance, elongation and termination. Elongation is thought to be a critical stage for the regulation of gene expression. ELL (11-19 lysine-rich leukemia protein), also designated MEN, functions as an RNA polymerase II elongation factor that increases the rate of transcription by suppressing transient pausing by RNA polymerase II. It is also thought to regulate cellular proliferation. ELLMore Product Information