Anti-ZMYND11 Rabbit Polyclonal Antibody (Cy5.5®)

Leverantör: Bioss
BS-7697R-CY5.5
BOSSBS-7697R-CY5.5EA 10450 SEK
BOSSBS-7697R-CY5.5
Anti-ZMYND11 Rabbit Polyclonal Antibody (Cy5.5®)
Antikroppar
Chromatin reader that specifically recognizes and binds histone H3.3 trimethylated at 'Lys-36' (H3.3K36me3) and regulates RNA polymerase II elongation. Does not bind other histone H3 subtypes (H3.1 or H3.2) (By similarity). Colocalizes with highly expressed genes and functions as a transcription corepressor by modulating RNA polymerase II at the elongation stage. Acts as a tumor-suppressor by repressing a transcriptional program essential for tumor cell growth.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZMYND11
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:More Product Information
Order Now

SPECIFIKATIONER

Antigensymbol ZMYND11
Environmentally Preferable
Antikroppsnamn Zinc Finger, MYND-type Containing 11
Antigen synonymer MRD30|BRAM1|BS69
Antikroppstyp Primär
Klonalitet Polyclonal
Konjugation Cy5.5®
Reaktivitet Human,Rat,Mouse
Värd Rabbit
Gen-ID 10771
Isotyp IgG
ImmunKemi Yes
Immunogen 540-570/602
Form liquid
Rening Purified by Protein A
Koncentration 1 μg/μl
Korsad adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Förvaringstemperatur Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR