Anti-WDR61 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverantör: Bioss
BS-6886R-FITC
BOSSBS-6886R-FITCEA 10450 SEK
BOSSBS-6886R-FITC
Anti-WDR61 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikroppar
WDR61 is a subunit of the human PAF and SKI complexes, which function in transcriptional regulation and are involved in events downstream of RNA synthesis, such as RNA surveillance (Zhu et al., 2005 [PubMed 16024656]).[supplied by OMIM, Mar 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WDR61
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR