Anti-BMPR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverantör: Bioss
BS-4237R-HRP
BOSSBS-4237R-HRPEA 10450 SEK
BOSSBS-4237R-HRP
Anti-BMPR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikroppar
On ligand binding, forms a receptor complex consisting of two type II and two type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators. Binds to BMP-7, BMP-2 and, less efficiently, BMP-4. Binding is weak but enhanced by the presence of type I receptors for BMPs.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: BMPR2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR