Anti-ERBB2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverantör: Bioss
BS-4004R-A488
BOSSBS-4004R-A488EA 10700 SEK
BOSSBS-4004R-A488
Anti-ERBB2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikroppar
Protein tyrosine kinase that is part of several cell surface receptor complexes, but that apparently needs a coreceptor for ligand binding. Essential component of a neuregulin-receptor complex, although neuregulins do not interact with it alone. GP30 is a potential ligand for this receptor. Regulates outgrowth and stabilization of peripheral microtubules (MTs). Upon ERBB2 activation, the MEMO1-RHOA-DIAPH1 signaling pathway elicits the phosphorylation and thus the inhibition of GSK3B at cell membrane. This prevents the phosphorylation of APC and CLASP2, allowing its association with the cellMore Product Information