Anti-CDK11B Rabbit Polyclonal Antibody
Leverantör: Bioss
BS-3999R
BOSSBS-3999REA 8425 SEK
BOSSBS-3999R
Anti-CDK11B Rabbit Polyclonal Antibody
Antikroppar
Summary: This gene encodes a member of the p34Cdc2 protein kinase family. p34Cdc2 kinase family members are known to be essential for eukaryotic cell cycle control. This gene is in close proximity to CDC2L2, a nearly identical gene in the same chromosomal region. The gene loci including this gene, CDC2L2, as well as metalloprotease MMP21/22, consist of two identical, tandemly linked genomic regions which are thought to be a part of the larger region that has been duplicated. This gene and CDC2L2 were shown to be deleted or altered frequently in neuroblastoma with amplified MYCN genes. TheMore Product Information