Anti-DTNB Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverantör: Bioss
BS-3548R-A680
BOSSBS-3548R-A680EA 10350 SEK
BOSSBS-3548R-A680
Anti-DTNB Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikroppar
Dystrobrevin beta is a component of the dystrophin-associated protein complex and interacts with dystrophin short form DP71, syntrophins SNTG1 and SNTG2. It localises to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. 4 isoforms produced by alternative splicing have been identified.

Type: Primary
Antigen: DTNB
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 261-370/627
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR