Anti-MTOR Rabbit Polyclonal Antibody (Cy5®)
Leverantör: Bioss
BS-3494R-CY5
BOSSBS-3494R-CY5EA 10800 SEK
BOSSBS-3494R-CY5
Anti-MTOR Rabbit Polyclonal Antibody (Cy5®)
Antikroppar
Serine/threonine protein kinase which is a central regulator of cellular metabolism, growth and survival in response to hormones, growth factors, nutrients, energy and stress signals. MTOR directly or indirectly regulates the phosphorylation of at least 800 proteins. Functions as part of 2 structurally and functionally distinct signaling complexes mTORC1 and mTORC2 (mTOR complex 1 and 2). Activated mTORC1 up-regulates protein synthesis by phosphorylating key regulators of mRNA translation and ribosome synthesis. This includes phosphorylation of EIF4EBP1 and release of its inhibition towardMore Product Information