Anti-CDK9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Leverantör: Bioss
BS-3097R-A647
BOSSBS-3097R-A647EA 10700 SEK
BOSSBS-3097R-A647
Anti-CDK9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikroppar
Protein kinase involved in the regulation of transcription. Member of the cyclin-dependent kinase pair (CDK9/cyclin-T) complex, also called positive transcription elongation factor b (P-TEFb), which facilitates the transition from abortive to productive elongation by phosphorylating the CTD (C-terminal domain) of the large subunit of RNA polymerase II (RNAP II) POLR2A, SUPT5H and RDBP. This complex is inactive when in the 7SK snRNP complex form. Phosphorylates EP300, MYOD1, RPB1/POLR2A and AR, and the negative elongation factors DSIF and NELF. Regulates cytokine inducible transcriptionMore Product Information