Anti-HERC3 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)

Leverantör: Bioss
BS-15461R-A488
BOSSBS-15461R-A488EA 10375 SEK
BOSSBS-15461R-A488
Anti-HERC3 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Antikroppar
HERC3 is a member the HERC ubiquitin ligase family. The encoded protein is located in the cytosol and binds ubiquitin via a HECT domain. Mutations in this gene have been associated with colorectal and gastric carcinomas. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.

Type: Primary
Antigen: HERC3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 401-500/1050
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR