Anti-SRG4 Rabbit Polyclonal Antibody

Leverantör: Bioss
BS-1220R
BOSSBS-1220REA 8425 SEK
BOSSBS-1220R
Anti-SRG4 Rabbit Polyclonal Antibody
Antikroppar
Testis and spermatogenesis related gene 4 may play a role in the development of cryptorchidism, the failure of the testis to descend from the abdomen into the scrotum [RGD] [TISSUE SPECIFICITY] Testis.

Recommended Dilutions: Western Blot: 1:100 to 1000; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.

Type: Primary
Antigen: SRG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR