Anti-SOX8 Rabbit Polyclonal Antibody (Cy3®)
Leverantör: Bioss
BS-11600R-CY3
BOSSBS-11600R-CY3EA 10450 SEK
BOSSBS-11600R-CY3
Anti-SOX8 Rabbit Polyclonal Antibody (Cy3®)
Antikroppar
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of thisMore Product Information