Anti-SOX6 Rabbit Polyclonal Antibody
Leverantör: Bioss
BS-11599R
BOSSBS-11599REA 8425 SEK
BOSSBS-11599R
Anti-SOX6 Rabbit Polyclonal Antibody
Antikroppar
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of thisMore Product Information