Anti-TXNDC11 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverantör: Bioss
BS-11439R-A680
BOSSBS-11439R-A680EA 10350 SEK
BOSSBS-11439R-A680
Anti-TXNDC11 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikroppar
May act as a redox regulator involved in DUOX proteins folding. The interaction with DUOX1 and DUOX2 suggest that it belongs to a multiprotein complex constituting the thyroid H₂O₂ generating system. It is however not sufficient to assist DUOX1 and DUOX2 in H₂O₂ generation.

Type: Primary
Antigen: TXNDC11
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 601-700/985
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR