Anti-HSP 40-4/HDJ2/DNAJA1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Leverantör: Bioss
BS-11277R-A555
BOSSBS-11277R-A555EA 10375 SEK
BOSSBS-11277R-A555
Anti-HSP 40-4/HDJ2/DNAJA1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikroppar
DnaJ-like proteins interact with HSP 70 molecular chaperones and function to facilitate protein folding and mitochondrial protein import. HSP 40-4, also known as HDJ2, is the human DnaJ homolog that functions as a co-chaperone with a cysteine-rich zinc finger domain. The cellular redox enzyme thioredoxin interacts with HSP 40-4, and oxidation and reduction reversibly regulate HSP 40-4 function in response to the changing redox states of the cell. The zinc finger domain of HSP 40-4 may act as a redox sensor of chaperone-mediated protein-folding machinery, since HSP 40-4 inactivation leads toMore Product Information