Anti-AP2M1 Rabbit Polyclonal Antibody (Cy7®)
Leverantör: Bioss
BS-11242R-CY7
BOSSBS-11242R-CY7EA 10800 SEK
BOSSBS-11242R-CY7
Anti-AP2M1 Rabbit Polyclonal Antibody (Cy7®)
Antikroppar
Adaptins are heterotetrameric subunits of adaptors, which are complexes involved in the formation of Clathrin-coated pits for vesicle-mediated endocytosis. Clathrin and its associated heterotetrameric protein complexes make up the main protein components of the coat surrounding the cytoplasmic face of coated vesicles. The Adaptin family, comprising a, b, and g classes, is also responsible for the transport of ligand-receptor complexes from plasma membranes and the trans-Golgi network to lysosomes. Two main types of adaptor proteins (APs), AP-1 and AP-2, are found in Clathrin-coatedMore Product Information