Anti-HIF1A Rabbit Polyclonal Antibody

Leverantör: Bioss
BS-0737R
BOSSBS-0737REA 8750 SEK
BOSSBS-0737R
Anti-HIF1A Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: HIF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR