Anti-CDX4 Mouse Monoclonal Antibody [clone: 1A11]

Leverantör: Abnova
H00001046-M07
ABNOH00001046-M07EA 4625 SEK
ABNOH00001046-M07
Anti-CDX4 Mouse Monoclonal Antibody [clone: 1A11]
Antikroppar
Type: Primary
Antigen: CDX4
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFIKATIONER

Antigensymbol CDX4
Environmentally Preferable
Antikroppsnamn Caudal type homeobox 4
Antigen synonymer Homeobox protein CDX-4.|CDX4_HUMAN|CDX4|Caudal-type homeobox protein 4
Antikroppstyp Primär
Klonalitet Monoclonal
Konjugation Unconjugated
Klon 1A11
Reaktivitet Human
Värd Mouse
Gen-ID 1046
Isotyp
Western blot Yes
ELISA Yes
Korsad adsorption No
Förvaringstemperatur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR