Anti-ALOX15 Mouse Monoclonal Antibody [clone: 1A11]

Leverantör: Abnova
H00000246-M07
ABNOH00000246-M07EA 4625 SEK
ABNOH00000246-M07
Anti-ALOX15 Mouse Monoclonal Antibody [clone: 1A11]
Antikroppar
Type: Primary
Antigen: ALOX15
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFIKATIONER

Antigensymbol ALOX15
Antikroppsnamn Arachidonate 15-lipoxygenase
Environmentally Preferable
Antigen synonymer LOG15|Arachidonate 15-lipoxygenase|Arachidonate omega-6 lipoxygenase|15-LOX|ALOX15
Antikroppstyp Primär
Klonalitet Monoclonal
Konjugation Unconjugated
Klon 1A11
Reaktivitet Human
Värd Mouse
Gen-ID 246
Isotyp
Western blot Yes
ELISA Yes
Korsad adsorption No
Förvaringstemperatur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR