Anti-GNRH1 Rabbit Polyclonal Antibody

Leverantör: Abgent
AP7754A
ABGEAP7754AEA 4125 SEK
ABGEAP7754A
Anti-GNRH1 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: GNRH1 (gonadotropin-releasing hormone 1 (luteinizing-releasing hormone))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR