Anti-RBM24 Rabbit Polyclonal Antibody

Leverantör: Abgent
AP5609A
ABGEAP5609AEA 4125 SEK
ABGEAP5609A
Anti-RBM24 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: RBM24 (RNA binding motif protein 24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR