Anti-BAD Rabbit Polyclonal Antibody

Leverantör: Abgent
AP3777O
ABGEAP3777OEA 4125 SEK
ABGEAP3777O
Anti-BAD Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: BAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR