Anti-ARRB1 Rabbit Polyclonal Antibody

Leverantör: Abgent
AP12005B
ABGEAP12005BEA 4125 SEK
ABGEAP12005B
Anti-ARRB1 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: ARRB1 (arrestin, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR