Antibody labelling kits, HisTag® Antibody HRP Conjugate kit
Leverantör: Merck Millipore (Novagen)
71840-3
71840-3EA 12175 SEK
71840-3
Antibody labelling kits, HisTag® Antibody HRP Conjugate kit
Analyser Proteinanalys
The HisTag® Antibody HRP Conjugate kit contains a horseradish peroxidase-conjugated HisTag® Monoclonal Antibody that is useful as a single detection reagent, thus eliminating cross-reactivity associated with secondary antibody reagents. It recognises five consecutive histidine residues regardless of the surrounding amino acid context. The purified antibody conjugate produces a strong signal at a 1:1000 to 1:2000 dilution on Western blots and dot blots. The HisTag® Antibody HRP Conjugate also detects the recombinant marker bands in the Novagen Perfect Protein™ Western Markers for convenient visualisation of accurate internal standards on Western blots.
Using standard HRP detection methods, as little as 1 ng protein can be detected on dot blots. Cross-reactivity to bacterial, insect, yeast or mammalian cell proteins is negligible. The package size provides enough conjugate for 25 to 50 blots and includes 5% alkali-soluble casein for the blocking reagent.