pH/mV/°C meter, handheld, Orion Star™ A321
Leverantör: Thermo Orion
STARA3210 STARA3215
662-1446EA 14300 SEK
662-1446 662-1447
pH/mV/°C meter, handheld, Orion Star™ A321
Elektrokemimätare pH-mätare
This portable pH meter is designed a wide range of pH, mV, ORP and temperature testing and field applications. The meter is suitable for use in the most demanding locations thanks to its IP 67-rated housing.
  • Up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading,
More Product Information