Anti-Glucose Transporter-3 Rabbit Polyclonal Antibody

Leverantör: Merck Millipore (Oncogene)
400062-50
400062-50EA 11525 SEK
400062-50
Anti-Glucose Transporter-3 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: Glucose Transporter-3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR