Anti-COL10 Rabbit Polyclonal Antibody

Leverantör: Merck Millipore (Oncogene)
234196-500
234196-500EA 12950 SEK
234196-500
Anti-COL10 Rabbit Polyclonal Antibody
Antikroppar
Type: Primary
Antigen: COL10 (collagen, type X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR