VWR®, VP 2 Autovac kemiskt resistent vakuumsystem
Leverantör: VACUUBRAND
20696767
181-0159EA 0 SEK
181-0159
VWR®, VP 2 Autovac kemiskt resistent vakuumsystem
Pumpar Membranpumpar
The VP 2 Autovac chemistry pumping unit, with vacuum controller, meets the demands of modern rotary evaporation. The hysteresis-free vacuum control prevents boiling retardation and foaming, therefore, ensuring consistently high process safety and an exceptionally high performance, even at low vacuum. This control enables detection of vapour pressure and fully automatic adjustment of the vacuum level to the process requirements without any user programming or intervention.
  • Exceptionell kemikalieresistens och överlägsen ångtolerans
  • Utmärkt slutvakuum även
More Product Information