TriLink® 2'-Deoxyguanosine-5'-O-(1-Thiotriphosphate)
Leverantör: TriLink BioTechnologies
N-8003-1 N-8003-10 N-8003-5
SOLUN-8003-1EA 875 SEK
SOLUN-8003-1 SOLUN-8003-10 SOLUN-8003-5
TriLink® 2'-Deoxyguanosine-5'-O-(1-Thiotriphosphate)
Nukleinsyrareagens Nukleotider
  • Grade: RUO
Observera: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=770e4fc24e1656ab41a4afb05b398747715bbe83&vid=770e4fc24e1656ab41a4afb05b398747715bbe83&prid=web&clid=SAPDAM