Human IL-1 alpha ELISA Kits
Leverantör: Abcam
AB46028-96 AB46028-2X96
ABCAAB46028-96EA 10775 SEK
ABCAAB46028-96 ABCAAB46028-2X96
Human IL-1 alpha ELISA Kits
Analyser ELISAs
Human IL-1 alpha ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human IL-1 alpha in Human in plasma, cell culture supernatant, Serum samples.